Learn More
MSC Mitutoyo Tipped Scriber, Carbide Tipped, 80 mm, 1 ea
Supplier:  MSC 95296984
Encompass
This carbide-tipped scriber is a replacement part designed for use with specific height gauges. Featuring a robust construction, this standard-type scriber is intended for marking and layout applications in various metalworking environments.
  • Carbide-tipped construction for durability
  • Compatible with a wide range of height gauges, including the 192, 514, and 570 series
  • Standard scriber design for reliable performance
  • Dimensions of 9 mm by 32 mm
Catalog No. 50-005-16059