Learn More
TARGETMOL CHEMICALS INC MTOR INHIBITOR 13 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T93105MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. mTOR inhibitor 13 is a selective mTOR inhibitor with IC50 of 0.29nM and 119nM for mTOR and PI3K(alpha) respectively. purity: 99%
Catalog No. 50-257-9008