Learn More
TARGETMOL CHEMICALS INC MTOR INHIBITOR 9A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T677055MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. mTOR inhibitor 9a inhibited the growth of human LNCap cells with an ic50 of 80 nm. 1-methyl-3 -{4-[4-(8-oxa-3-azabicyclo[3.2.1]oct-3-yl)thieno[32-d]pyrimidin-2-yl]phenyl}urea may have antitumor activity. purity: 99%
Catalog No. 50-257-9280