Learn More
Medchemexpress LLC mTOR inhibitor WYE-28 | 1062172-60-4 | MFCD16661023 | 99.3% | 586.64 g·mol⁻1 | C30H34N8O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1187171MG
Encompass_Preferred
mTOR inhibitor WYE-28 is a potent, selective small-molecule inhibitor of mTOR used for cell signaling and oncology research. It demonstrates sub-nanomolar mTOR inhibition and is supplied as a high-purity solid in milligram quantities with recommended storage conditions for research use.
  • Sub-nanomolar mTOR inhibition (IC50 = 0.08 nM) demonstrated in biochemical assays.
  • Additional activity against PI3Kα (IC50 = 6 nM), useful for pathway profiling.
  • High purity solid, suitable for cell-based assays and mechanistic studies.
  • Available in small milligram sizes for early-stage research and screening.
  • Powder storage: -20°C (up to 3 years); stock solutions: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-29580