Learn More
Medchemexpress LLC MTR-106 | 1639357-93-9 | 99.4% | 525.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1489535MG
Encompass_Preferred
MTR-106 is a potent and orally active G-quadruplex stabilizer and RNA polymerase I inhibitor. It induces apoptosis and inhibits cell growth, making it suitable for cancer research.
  • Potent and orally active G-quadruplex stabilizer
  • RNA polymerase I inhibitor
  • Induces apoptosis and inhibits cell growth
  • Has antitumor activity in both HR-deficient cells and PARPi-resistant cells
  • Induces cell cycle arrest and DNA damage
  • Suppresses tumor growth in BRCA-deficient and PARPi-resistant xenografts in nude mice
Catalog No. 50-003-60278