Learn More
Sigma Aldrich Fine Chemicals Biosciences mTRAQR Reagent Delta0 50 U
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 44400151KT
Encompass
Contains mTRAQ(R) Reagents Delta0 for 50 mTRAQ assays. One unit of reagent can label up to 100 mug of protein. Pack has 1 tube each of Delta0 containing 50 units of reagent. Optional Multiplex Buffer kit (Cat. No. 4381664) to be ordered separately.
Catalog No. 50-290-7959