Learn More
Cayman Chemical mtrmLST8Raptr Complexhu 10ug
SDP
Supplier:  Cayman Chemical 3398510 UG
Encompass_Preferred
Source: Active recombinant human N-terminal DYKDDDDK-tagged mTOR, N-terminal His-tagged mLST8, and N-terminal His-tagged Raptor expressed in insect cells • Amino acids: 1,362-2,549 (mTOR), 2-326 (mLST8), and 2-1,335 (Raptor) • MW: 137 (mTOR), 37 (mLST8), and 150 (Raptor) kDa
Catalog No. 50-295-0305