Learn More
Biotium MTS-Tetramethylrhodamine (MTS-TAMRA)
Supplier:  Biotium 91030
Encompass_Preferred
MTS-TAMRA (MTS-5(6)-carboxytetramethylrhodamine, mixed isomers) is a fast-reacting and highly selective thiol-reactive label that permits one to attach the popular fluorescent dye carboxytetramethylrhodamine to a thiol group. Properties: Ex/Em (MeOH) = 540/565 nm; Red solid soluble in DMF or DMSO; Store at -20°C and protect from light; C28H29N3O6S2; MW: 567.
Catalog No. 50-196-4831