Learn More
Biotium MTSEA (FULL CHEMICAL NAME: (2-AMINOETHY
Supplier:  Biotium 91022
Encompass_Preferred
MTSEA (2-Aminoethyl methanethiosulfonate hydrobromide) is a positively charged MTS reagent. Off-white solid soluble in water, DMSO or DMF Store desiccated at -20°C C3H10BrNO2S2 MW: 236.15
Catalog No. NC0903425