Learn More
Cayman Chemical MTSETchlorIde 5mg
SDP
Supplier:  Cayman Chemical 210695 MG
Encompass_Preferred
A positively charged sulfhydryl-specific reagent that reacts with substituted-cysteines; used to provide functional information about relative positions of amino acids within a protein and to probe binding site electrostatic interactions
Catalog No. 50-298-3482