Learn More
Medchemexpress LLC MTSS1 protein, human (His) | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7593010UG
Recombinant human MTSS1 (metastasis suppressor 1) is supplied as an N-terminal 6xHis-tagged protein expressed in Escherichia coli and purified for research applications. It is provided at high purity (>95% by reducing SDS-PAGE) and includes sequence information corresponding to residues M1-S250, making it suitable for biochemical assays, antibody validation, and protein-protein interaction studies.
- N-terminal 6xHis tag for affinity purification and detection.
- Expression host: Escherichia coli.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Sequence corresponds to residues M1-S250 (Protein ID EAW92073).
- Available in small research-scale pack sizes, including 10 μg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.