Learn More
Medchemexpress LLC MTSSL | 81213-52-7 | 95.07% | 25 MG

Supplier: Medchemexpress LLC 25MGHY130509
MTSSL (Otmpmms) is a highly reactive thiol-specific spin label. It is utilized for labeling protein thiol residues to determine protein structure and dynamics, and for studies involving protein-protein interactions.
- Highly reactive thiol-specific spin label
- Labels protein thiol residues for structure and dynamics determination
- Used in protein-protein interaction studies
- Solid appearance
- Off-white to light yellow color
- Molecular weight: 264.38
- Chemical formula: C10H18NO3S2
- Purity: 95.07%
- Recommended storage temperature: 4°C, stored under nitrogen
- In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.