Learn More
Gold Biotechnology Inc Thiazolyl Blue Tetrazolium Bromide (MTT) 5 g
SDP
Supplier:  Gold Biotechnology Inc T0305
Encompass_Preferred
Thiazolyl blue tetrazolium bromide (often called MTT) is a common dye used in Cell Proliferation assays or Cell Growth assays. MTT is converted to water-insoluble, dark blue MTT-formazan by mitochondrial dehydrogenases of living cells. The intensity of the color can then be measured colorimetrically at a wavelength of 570 nm.
Catalog No. NC2943381