Learn More
Research Products International Corp MTT [Thiazolyl Blue Tetrazolium Bromide], 1 Gram
SDP
Supplier:  Research Products International Corp M920501.0
Encompass_Preferred
Water soluble salt of Tetrazolium. Yields a yellowish solution when prepared in media or salt solutions lacking phenol red. By cleavage of the tetrazolium ring by dehydrogenase enzymes, dissolved MTT is converted into insoluble purple formazan.
Catalog No. 50-488-833