Learn More
ABclonal Technology MTTP Rabbit pAb
SDP
Supplier:  ABclonal Technology A14028500UL
Encompass_Preferred
MTP encodes the large subunit of the heterodimeric microsomal triglyceride transfer protein. Protein disulfide isomerase (PDI) completes the heterodimeric microsomal triglyceride transfer protein, which has been shown to play a central role in lipoprotein assembly. Mutations in MTP can cause abetalipoproteinemia.
Catalog No. 50-001-06723