Learn More
Medchemexpress LLC Mtx-23 (protac AR-V7 degrader-2) | 2488296-74-6 | 99.79% | 882.05 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1487711MG
Encompass_Preferred
MTX-23 is an AR-based PROTAC that inhibits CaP cellular proliferation by degrading AR-V7 and AR-FL. It induces apoptosis, offering a potential therapeutic approach for androgen receptor-related cancers.
  • AR-based PROTAC
  • Inhibits CaP cellular proliferation
  • Degrades AR-V7 and AR-FL
  • Induces apoptosis
Catalog No. 50-003-61375