Learn More
Selleck Chemical LLC MTX-531 E4680-100MG
SDP
Supplier:  Selleck Chemical LLC E4680100MG
Encompass_Preferred
MTX-531(NSC827271) is a first-in-class potent and selective inhibitor of EGFR and PI3K with an IC50 of 14 7 nM and 6 4 nM respectively It may play a role in treating Head and Neck Squamous Cell Carcinoma (HSNCC) squamous lung cancers and certain EGFR/PI3K-driven triple-negative breast cancers
Catalog No. 50-000-39450