Learn More
Genetex Inc MUC1 Monoclonal Antibody [MSVA-672R] HistoMAX™ (GTX04397) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04397
Encompass_Preferred
The MUC1 antibody [MSVA-672R] HistoMAX™ (GTX04397) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human epithelioid malignant mesothelioma tissue using GTX04397 MUC1 antibody [MSVA-672R] HistoMAX. Malignant mesothelioma epitheloid variant with strong predominantly membranous MUC1 immunostaining of tumor cells.
Catalog No. NC3666760