Learn More
Sino Biological Anti-MUC-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12123T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MUC1 (rh MUC1; NP_001018016.1; aa1-167). MUC1 specific IgG was purified by Human MUC1 affinity chromatography.
Catalog No. 50-228-3488