Learn More
Medchemexpress LLC MUC5AC motif peptide | 244049-41-0 | 99.0% | 1501.62 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP028010MG
Encompass_Preferred
MUC5AC motif peptide is a 16-amino acid fragment of mucin 5. This peptide mimics the human mucin tandem repeat of MUC5AC.
  • For research use only
  • Sequence: Gly-Thr-Thr-Pro-Ser-Pro-Val-Pro-Thr-Thr-Ser-Thr-Thr-Ser-Ala-Pro
  • Shortened sequence: GTTPSPVPTTSTTSAP
  • Appears as a white to off-white solid
Catalog No. 50-004-24965