Learn More
Selleck Chemical LLC Mucic Acid S3964-25mg
SDP
Supplier:  Selleck Chemical LLC S396425MG
Encompass_Preferred
Mucic Acid (Schleimsaure Saccharolactic acid Tetrahydroxyadipic acid D-Glucaric acid Galactaric acid) is the dicarboxylic sugar acid resulting from oxidation glactose with dilute nitric acid It is a substrate of galactarate O-hydroxycinnamoyltransferase
Catalog No. 50-310-9544