Learn More
Medchemexpress LLC Mucic acid (galactaric acid) | 526-99-8 | MFCD00004239 | 97.0% | 210.14 g/mol | C6H10O8 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01441010MM/1ML
Encompass_Preferred
Mucic acid (galactaric acid) is an aldaric diacid used as a research reagent and polymer building block. It contains multiple hydroxyl and carboxyl groups that enable derivatization and crosslinking for synthesis of functional polymers and biomaterials. It has been used in studies of bone tissue engineering where it can influence osteogenic markers. Verify solvent and purity for your intended application.
  • Hexaric acid with formula C6H10O8 and molecular weight 210.14 g/mol.
  • Useful building block for polymer synthesis and functionalization.
  • Reported to stimulate Runx2 mRNA expression in bone-related research.
  • Prepared solution formats are available for convenient handling in assays.
  • Verify lot-specific purity and solvent from the supplier certificate of analysis.
Catalog No. 50-003-03940