Learn More
Medchemexpress LLC Mucic acid (galactaric acid) | 526-99-8 | MFCD00004239 | 99.4% | 210.14 g·mol⁻¹ | C6H10O8 | 250 G

Supplier: Medchemexpress LLC 250GHYW014410
Mucic acid (galactaric acid) is a sugar-derived dicarboxylic acid used as a building block in polymer synthesis and in bone tissue engineering research. It is supplied as a white to off-white solid with high purity for laboratory applications.
- High purity (~99.4%) suitable for laboratory research.
- White to off-white solid, easy to handle and store.
- Chemical formula C6H10O8; molar mass 210.14 g·mol⁻¹.
- Synonym galactaric acid; derived from sugar feedstocks.
- Useful as a building block for polymer synthesis and osteogenic studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.