Learn More
Creative Biomart Mucin-1 MUC1
Supplier:  Creative Biomart THP0275
Encompass
Human Mucin-1 Llama IgG2b Fc Tag low endotoxin is expressed from human 293 cells (HEK293) It contains AA Ser 33 - Gly 167 Predicted N-terminus Ser 33 This protein carries a llama IgG2b Fc tag at the C-terminus The protein has a calculated MW of 42 4 kDa The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation
Catalog No. 50-003-32703