Learn More
Biocare Medical, LLC Mucin 5AC Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM231A
Encompass_Preferred
Mucins are high molecular weight glycoproteins with 80% carbohydrate content and the remaining 20% consisting of a protein core. Mucin 5AC antibody (MUC5AC) is defined as a secretory-type mucin and is seen mainly in gastric foveolar cells. A study has suggested that MUC5AC expression is an early event in tumorigenesis. Another study indicates up-regulation of MUC5AC may be associated with carcinogenesis, malignant potential, progression and clinical behaviors in colorectal signet-ring cell carcinoma.
Catalog No. 50-826-17