Learn More
Genetex Inc Mucin 5AC Monoclonal Antibody [MSVA-109M] HistoMAX™ (GTX04364) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04364
Encompass_Preferred
"The Mucin 5AC antibody [MSVA-109M] HistoMAX™ (GTX04364) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human gastric adenocarcinoma (100% of tumor cells) tissue using GTX04364 Mucin 5AC antibody [MSVA-109M] HistoMAX. Strong MUC5AC immunostaining in an adenocarcinoma of the stomach 100% of tumor cells."
Catalog No. NC3663012