Learn More
Sigma Aldrich Fine Chemicals Biosciences Mucin from porcine stomach, 84082-64-4, MFCD00131629, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M2378100G
Encompass
Mucin from porcine stomach has been used: in the standard curve generation for quantifying mucin-2 generated during mucosal inflammation in mice, as a component of digestion fluid solutions in in vitro human digestion model, and as a component of simulated saliva fluid. Mucin from porcine stomach was used in studies on the binding site of the galactose-specific agglutinin PA-IL from Pseudomonas aeruginosa.
Catalog No. NC1105098