Learn More
Medchemexpress LLC Mudanpioside J | 262350-52-7 | 1 MG

Supplier: Medchemexpress LLC HYN72781MG
Mudanpioside J is a monoterpene glycoside, identified as a metabolite of cortex moutan. It possesses a molecular formula of C31H34O14 and a molecular weight of 630.59. This compound is typically provided for research purposes only.
- Monoterpene glycoside compound
- Metabolite of cortex moutan
- Molecular formula: C31H34O14
- Molecular weight: 630.59
- High purity: 95.0%
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.