Learn More
Teknova Mueller Hinton Agar Plates, 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Supplier:  Teknova M00274PK
Encompass
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.
Catalog No. 50-238-7465