Learn More
Medchemexpress LLC Mulberroside A | 102841-42-9 | 568.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN061910MM/1ML
Encompass_Preferred
Mulberroside A is a primary bioactive constituent found in mulberry (Morus alba L.). It has anti-inflammatory and antiapoptotic effects by decreasing the expressions of TNF-α, IL-1β, and IL-6, and inhibiting the activation of NALP3, caspase-1, and NF-κB, as well as the phosphorylation of ERK, JNK, and p38. It also exhibits inhibitory activity against mushroom tyrosinase.
  • Anti-inflammatory and antiapoptotic properties.
  • Inhibits activation of NALP3, caspase-1, and NF-κB.
  • Decreases expression of TNF-α, IL-1β, and IL-6.
  • Exhibits inhibitory activity against mushroom tyrosinase.
  • Can be used in in vitro and in vivo studies.
Catalog No. 50-003-88361