Learn More
Medchemexpress LLC Mulberroside A | 102841-42-9 | 568.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0619100MG
Encompass_Preferred
Mulberroside A is a primary bioactive constituent found in mulberry (Morus alba L.). It has been shown to decrease the expressions of TNF-α, IL-1β, and IL-6, and inhibits the activation of NALP3, caspase-1, and NF-κB, as well as the phosphorylation of ERK, JNK, and p38. These actions demonstrate its anti-inflammatory and antiapoptotic effects. Mulberroside A also exhibits inhibitory activity against mushroom tyrosinase with an IC50 of 53.6 μM.
  • Decreases the expressions of TNF-α, IL-1β, and IL-6.
  • Inhibits the activation of NALP3, caspase-1, and NF-κB.
  • Inhibits the phosphorylation of ERK, JNK, and p38.
  • Exhibits anti-inflammatory and antiapoptotic effects.
  • Shows inhibitory activity against mushroom tyrosinase.
Catalog No. 50-003-88951