Learn More
Apexbio Technology LLC Mulberroside C 102841-43-0 20mg
SDP
Supplier:  Apexbio Technology LLC N251920
Encompass_Preferred
Mulberroside C (CAS 102841-43-0) is a small-molecule inhibitor designed to suppress hepatitis C virus (HCV) replication Mulberroside C exerts its biological activity primarily through inhibition of the HCV replication cycle In cell-based in vitro assays Mulberroside C demonstrates antiviral activity exhibiting an IC50 value within the micromolar range Based on these pharmacological properties Mulberroside C holds research potential in antiviral screening HCV pharmacological studies and exploration of viral replication inhibitor compounds
Catalog No. 50-000-94393