Learn More
Medchemexpress LLC Mulberroside F | 193483-95-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN35181MG
Encompass_Preferred
Mulberroside F is one of the main bioactive constituents found in mulberry (Morus alba L.). It demonstrates inhibitory effects on tyrosinase activity and melanin formation.
  • Inhibits tyrosinase activity.
  • Inhibits melanin formation.
  • Exhibits superoxide scavenging activity.
  • Protects against auto-oxidation.
Catalog No. 50-004-19024