Learn More
Selleck Chemical LLC Mulberry bark Extract E3595-5MG
SDP
Supplier:  Selleck Chemical LLC E35955MG
Encompass_Preferred
White Mulberry Root-Bark Extract is extracted from the root-bark of Morus alba which can regulate multiple metabolic pathways to exert hypoglycemic effects
Catalog No. 50-000-39777