Learn More
SPEX CERTIPREP LLC 3P ICP-MS Multi-Component 5 Component EPA Method 200.8 Internal Standard at 100 g/mL in 2% HNO3
Supplier:  SPEX CERTIPREP LLC 3P ICPMSIS2100
Encompass_Preferred
(HPS by Zeptometrix) ICP-MS Multi-Component 5 Component EPA Method 200.8 Internal Standard at 100 g/mL in 2% HNO3. This standard is traceable to NIST SRM 3100 series. ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited. 12 month expiration date. Components: Bismuth, Indium, Scandium, Terbium, Yttriumm.
Catalog No. NC2340130