Learn More
GENIE GROUP EMF Field Meter; Pocket; Simultaneous Magnetic; Electric; RF Strenght; Color TFT
Supplier:  GENIE GROUP EMF450
Encompass_Preferred
Multi-Field measurements: Magnetic Field, ElectricField, and Radio Frequency (RF) Strength• Magnetic Field Triple Axis Sensor (X, Y, Z)• Audible alarm• 2.4" (240 x 320 pixels) color TFT display• Adjustable screen brightness (low, medium, high)• Store up to 20 RF historical records• Data Hold and Auto Power Off with disable feature• Five languages: English, Traditional/SimplifiedChinese, Japanese, Spanish• Complete with 3 AAA batteries
Catalog No. NC1190319