Learn More
Medchemexpress LLC Multi-Leu peptide (triacetate) | 98.64% | 1231.53 | 50MG
SDP
Supplier:  Medchemexpress LLC HYP10422A50MG
Encompass_Preferred
Multi-Leu peptide (triacetate) is a potent inhibitor of PACE4 with a Ki of 22 nM. It competitively binds to the active site of PACE4, thereby inhibiting its catalytic activity. This product is suitable for studying the specific mechanism of PACE4 in the development of prostate cancer.
  • Potent inhibitor of PACE4
  • Competitively binds to PACE4 active site
  • Inhibits catalytic activity of PACE4
  • Useful for studying PACE4 in prostate cancer development
Catalog No. 50-004-23214