Learn More
Research Products International Corp MultiMagstir Genie, Four Position Magnetic Stirrer
SDP
Supplier:  Research Products International Corp 150604
Encompass
The revolutionary MagStir Genie provides low to high speed stirring and editable memory profiles. Microprocessor controls offer precision and reproducibility without internal heat build-up.
  • Speed range of 10-1800 RPM including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers.
  • May be used with any regular flask/beaker or spinner flask.
  • Three power levels available for various sample viscosities.
  • Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction.
  • Manual mode provides stir, stir reverse, and time functions.
  • Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to 9 distinct steps.
  • Two year warranty.
Catalog No. 50-999-091