Learn More
Molecular Devices Molecular Devices SpectraMax Mini Microplate Reader, Absorbance, Fluorescence, Luminescence
Supplier:  Molecular Devices SMAXMINIAFL
Encompass_Preferred
The SpectraMax Mini is a compact benchtop multi-mode microplate reader designed for routine absorbance, fluorescence intensity, and luminescence assays across 6-384 well plates. It supports endpoint, kinetic, and spectral scanning measurements, offers temperature control and multiple shaking modes, and integrates with data acquisition software for automated workflows.
  • Compact footprint for limited bench space.
  • Supports 6, 12, 24, 48, 96, and 384-well plates.
  • Measurement modes: absorbance, fluorescence intensity, luminescence.
  • Wavelength coverage for absorbance (200-1000 nm) and fluorescence (250-850 nm).
  • Temperature control from 4°C above ambient to 45°C with ±1°C uniformity at 37°C.
  • Linear, orbital, and double orbital shaking options.
  • Endpoint, kinetic, and spectral scanning read modes.
  • Robotic compatible for high-throughput workflows.
Catalog No. 50-000-14380
Add to Cart
Request a Quote