Learn More
Amsbio LLC Multi-normal human tissue arra
Supplier:  Amsbio LLC FDA999Y2
Encompass
Multi-normal human tissue array rerpresenting FDA guideline for antibody cross-reactivity testing 33 types of human organs 99 cores each type taken from 3 individuals (core size 1.5mm).
Catalog No. 50-256-4468