Learn More
Hach Company Multi-purpose half cable SD900
Supplier:  Hach Company 8756900
Encompass
Multi-purpose half cable 10 ft 7 Pin Auxiliary connector on one end open leads on the other end Connects an SD900 sampler to a non-Sigma flow meter
Catalog No. 50-002-07098