Learn More
Hach Company Multi-purpose half cable SD900
Supplier:  Hach Company 8756800
Encompass
Multi-purpose half cable 25 ft 7 Pin Auxiliary connector on one end open leads on the other end Connects SD900 to Sigma flow meter
Catalog No. 50-002-07212