Learn More
Med Vet International Exel Multi Sample Needle Holder, 100/Box
Supplier:  Med Vet International 26530EX
Encompass_Preferred
This Exel International 26530EX multi sample needle holder contains no latex and has sharp and smooth edges to make penetration painless for patiients and provides easy connection to rubber stoppers.
Catalog No. 50-264-3637