Learn More
Research Products International Corp Multi Vortex-Genie
SDP
Supplier:  Research Products International Corp 155582
Encompass
The Multi Vortex-Genie provides orbital and vortex mixing of liquids in a variety of container sizes from micro-tubes up to 250ml flasks. Electronic speed control ramps up gently to selected speed to prevent spillage from open containers. Simply place tubes or container in the rack, set speed, and time. Compact space saving design fits easily on a benchtop and may be used in a cold room or incubator. Racks are included and no additional holders are required.
Catalog No. 50-197-6853