Learn More
Medchemexpress LLC HY-B2140 5mg Medchemexpress, Multifungin CAS:3679-64-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB21405MG
Encompass_Preferred
Medchemexpress, HY-B2140 5mg Multifungin CAS:3679-64-9 Multifungin (Bromochlorosalicylanilide) is an antifungal that treats oral candidiasis. Multifungin prevents the formation and accumulation of Zearalenone and reduces the fungal population in stored-crushed corn. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0176