Learn More
LABSYSTEMS DIAGNOSTICS OY Multilevel Control in DBS format
Supplier:  LABSYSTEMS DIAGNOSTICS OY 710012001
Encompass
NeoMass AAAC Plus DBS controls, are intended to be used for internal quality assurance in clinical laboratories that screen newborns for inborn errors of metabolism using a tandem mass spectrometer (MS/MS).NeoMass AAAC Plus DBS Controls are prepared by using human blood and are available as a controls set with 3 levels i.e., low, medium and high.
Catalog No. NC2865472