Learn More
Cole-Parmer Fluke 115 Digital Multimeter with True-RMS
Supplier:  Cole-Parmer 2601630
Encompass
This compact meter features True-RMS AC and DC voltage, 6000-count resolution, min/max/average recording capabilities, frequency, auto/manual ranging, resistance, continuity/diode test, capacitance, display hold, white LED backlit display, and accuracies of ±0.5%. It's ideal use is for general electrical, HVAC, utility, and field service applications.
Catalog No. 50-992-383