Learn More
Cole-Parmer Fluke 83-5 High Accuracy Digital Multimeter
Supplier:  Cole-Parmer 2608302
Encompass
Available for sale in the United States and Canada only The Fluke 83V industrial multimeter features a 6000-count, 3-3/4" digit display with a 32 segment bar graph. The 83V is CAT III 1000 V and CAT IV 600 V rated and can withstand 8000 volt spikes caused by load switching on industrial circuits. Meter is auto and manual ranging for maximum flexibility during testing. Meter provides Min/Max and average readings allowing you to capture variations.
Catalog No. 50-993-340