Learn More
Amsbio LLC Multiple organ tumor tissue ar
Supplier:  Amsbio LLC MC2082C
Encompass
Multiple organ tumor tissue array including pathology grade TNM and clinical stage 192 cases/192 cores replacing MC2082b
Catalog No. 50-256-4166