Learn More
RAE Systems Inc MULTIRAE LITE PUMPED/10.6 EV
SDP
Supplier:  RAE Systems Inc MAB3A2C112E02M/DEL
Encompass_Preferred
MultiRAE Lite Pumped / 10.6 eV PID / LEL / H2S / CO / O2 / Li-ion / Non-Wireless
Catalog No. NC1425387